Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
This target displays homology in the following species: Cow: 86%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 92%; Pig: 85%; Rabbit: 93%; Rat: 92%; Zebrafish: 77%
Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hNaDC3; membrane transporter responsible for the transport of tricarboxylic acid cycle intermediates; Na(+)-coupled carboxylate transporter 3; Na(+)/dicarboxylate cotransporter 3; NaC3; NaDC-3; NADC3; integral membrane transporter; SLC13A3; sodium-dependent high affinity dicarboxylate transporter 3; Sodium-dependent high-affinity dicarboxylate transporter 2; Solute carrier family 13 member 3; unnamed protein product
Gene Aliases: ARLIAK; NaC3; NADC3; SDCT2; SLC13A3
UniProt ID: (Human) Q8WWT9
Entrez Gene ID: (Human) 64849
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support