Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
AASDH encodes a member of the non-ribosome peptide syntesase (NRPS) enzyme family. The encoded protein contains an AMP-binding domain, PP-binding (phosphopantetheine, or pantetheine 4'phosphate-binding) domain and the Pyrrolo-quinoline quinon (PQQ) binding domain. The protein is expressed in several adult tissues. Covalently binds beta-alanine in an ATP-dependent manner to form a thioester bond with its phosphopantetheine group and transfers it to an, as yet, unknown acceptor. May be required for a post-translational protein modification or for post-transcriptional modification of an RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-aminoadipic 6-semialdehyde dehydrogenase; 2-aminoadipic 6-semialdehyde dehydrogenase antibody; AASDH antibody; ACSF4 antibody; Acyl-CoA synthetase family member 4; acyl-CoA synthetase family member 4 antibody; aminoadipate-semialdehyde dehydrogenase antibody; Beta-alanine-activating enzyme; LYS2 antibody; non-ribosomal peptide synthetase 1098; non-ribosomal peptide synthetase 1098 antibody; non-ribosomal peptide synthetase 998; non-ribosomal peptide synthetase 998 antibody; NRPS1098 antibody; NRPS998 antibody; Protein NRPS998; unnamed protein product
Gene Aliases: AASDH; ACSF4; HSPC318; LYS2; NRPS1098; NRPS998; U26
UniProt ID: (Human) Q4L235
Entrez Gene ID: (Human) 132949
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support