Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%
CRNKL1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: crn; Crn, crooked neck-like 1; Crooked neck homolog; crooked neck pre-mRNA splicing factor-like 1; crooked neck protein; crooked neck-like 1; Crooked neck-like protein 1; hCrn; SYF3 pre-mRNA-splicing factor; unnamed protein product
Gene Aliases: CGI-201; CLF; Clf1; CRN; CRNKL1; HCRN; MSTP021; SYF3
UniProt ID: (Human) Q9BZJ0
Entrez Gene ID: (Human) 51340
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support