Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Positive Control: K562
Predicted Reactivity: Chimpanzee (99%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The gamma globin genes (HBG1 and HBG2) are normally expressed in the fetal liver, spleen and bone marrow. Two gamma chains together with two alpha chains constitute fetal hemoglobin (HbF) which is normally replaced by adult hemoglobin (HbA) at birth. In some beta-thalassemias and related conditions, gamma chain production continues into adulthood. The two types of gamma chains differ at residue 136 where glycine is found in the G-gamma product (HBG2) and alanine is found in the A-gamma product (HBG1). The former is predominant at birth. The order of the genes in the beta-globin cluster is: 5'-epsilon - gamma-G - gamma-A - delta - beta-3'.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-gamma globin; epsilon 3 globin; fetal hemoglobin F subunit gamma 1; gamma A hemoglobin; gamma globin; Gamma-1-globin; Hb F Agamma; Hemoglobin gamma-1 chain; Hemoglobin gamma-A chain; Hemoglobin subunit gamma-1; hemoglobin Y beta-like embryonic chain; hemoglobin Y, beta-like embryonic chain; hemoglobin, epsilon 1; hemoglobin, gamma A; hemoglobin, gamma, regulator of; unnamed protein product
Gene Aliases: Hbb-y; Hbe1; HBG-T2; HBG1; HBGA; HBGR; HSGGL1; PRO2979
UniProt ID: (Human) P69891
Entrez Gene ID: (Human) 3047, (Rat) 94164
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support