Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Control: HeLa, HeLa membrane extract
Predicted Reactivity: Mouse (100%), Rat (98%), Chicken (96%), Bovine (99%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RAB5B play a role in protein transport. RAB5B is probably involved in vesicular traffic.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAB5B; Ras-related protein Rab-5B; unnamed protein product
Gene Aliases: RAB5B
UniProt ID: (Human) P61020
Entrez Gene ID: (Human) 5869
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support