Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • Laminin gamma-1,2,3 Antibodies

          Invitrogen

          Laminin gamma-1,2,3 Polyclonal Antibody

          Advanced Verification
          View all (2) Laminin gamma-1,2,3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Laminin gamma-1,2,3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          • Advanced Verification (1)
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Laminin gamma-1,2,3 Antibody (PA5-79576) in IHC (P)

          Immunohistochemistry (Paraffin) analysis of Laminin gamma-1,2,3 in paraffin-embedded section of mouse skeletal muscle tissue using Laminin gamma-1,2,3 Polyclonal Antibody (Product # PA5-79576). Heat mediated antigen retrieval was performed in EDTA buffer (pH 8.0, epitope retrieval solution). The tissue section was blocked with 10% goat serum. The tissue section was then incubated with the primary antibody a... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Western Blot (WB)
          Laminin gamma-1,2,3 Antibody in Western Blot (WB)
          Laminin gamma-1,2,3 Antibody
          Laminin gamma-1,2,3 Polyclonal Antibody

          Product Details

          PA5-79576

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          0.5-1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide mixture of laminin gamma1,2,3 (NKLNEIEGSLNKAKDEMKAS; DLEERVRRQRNHLHLLETSI; LQLDSHGALHHKLRQLEEES).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 4mg trehalose

          Contains

          0.05mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746691

          Product Specific Information

          This antibody reacts will all 3 isoforms of Laminin gamma: Laminin gamma-1, gamma-2 and gamma-3.

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: human A431 whole cell, human A549 whole cell, human PC-3 whole cell, human CACO-2 whole cell, rat NRK whole cell, rat RH35 whole cell, mouse NIH/3T3 whole cell, mouse HEPA1-6 whole cell. IHC: mouse heart tissue, mouse kidney tissue, rat cardiac muscle tissue, rat kidney tissue.

          Target Information

          Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: basement membrane protein; BM600-100kDa; Cell-scattering factor 140 kDa subunit; CSF 140 kDa subunit; Epiligrin subunit gamma; Kalinin subunit gamma; Kalinin/nicein/epiligrin 100 kDa subunit; Ladsin 140 kDa subunit; lamimin, gamma 2; Laminin B2 chain; laminin B2 precursor; Laminin B2t chain; laminin gamma2 chain; Laminin subunit gamma-1; Laminin subunit gamma-2; Laminin subunit gamma-3; laminin, gamma 1 (formerly LAMB2); laminin, gamma 2; laminin, gamma 3; Laminin-1 subunit gamma; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit gamma; Laminin-14 subunit gamma; Laminin-15 subunit gamma; Laminin-2 subunit gamma; Laminin-3 subunit gamma; Laminin-4 subunit gamma; Laminin-5 subunit gamma; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit gamma; Laminin-9 subunit gamma; Large adhesive scatter factor 140 kDa subunit; Nicein subunit gamma; nicein, 100kD; nicein, 100kDa; S-LAM gamma; S-laminin subunit gamma; unnamed protein product

          View more View less

          Gene Aliases: AA589349; AI562206; AW240805; B2e; B2T; BM600; CSF; EBR2; EBR2A; JEB3A; JEB3B; LAMB2; LAMB2T; LAMC1; LAMC2; LAMC3; LAMNB2; OCCM

          View more View less

          UniProt ID: (Human) P11047, (Human) Q13753, (Human) Q9Y6N6, (Mouse) Q9R0B6

          View more View less

          Entrez Gene ID: (Human) 3915, (Human) 3918, (Human) 10319, (Mouse) 226519, (Rat) 117036, (Mouse) 16782, (Rat) 192362, (Rat) 311862, (Mouse) 23928

          View more View less

          Function(s)
          extracellular matrix structural constituent extracellular matrix constituent conferring elasticity heparin binding structural molecule activity protein binding glycosphingolipid binding molecular_function extracellular matrix protein DNA metabolism protein
          Process(es)
          cell adhesion endoderm development tissue development cell migration extracellular matrix disassembly hemidesmosome assembly substrate adhesion-dependent cell spreading maintenance of blood-brain barrier positive regulation of cell adhesion positive regulation of epithelial cell proliferation positive regulation of muscle cell differentiation protein-containing complex assembly regulation of basement membrane organization positive regulation of integrin-mediated signaling pathway positive regulation of cell proliferation epidermis development positive regulation of cell migration cell morphogenesis visual perception astrocyte development retina development in camera-type eye protein complex assembly neuron projection development biological_process cell morphogenesis involved in differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline