Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PMCA4 ATPase Antibodies

          Invitrogen

          PMCA4 ATPase Polyclonal Antibody

          Advanced Verification
          View all (7) PMCA4 ATPase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PMCA4 ATPase Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Advanced Verification (1)
          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PMCA4 ATPase Antibody (PA5-83500) in ICC/IF

          Immunofluorecent analysis of PMCA4 ATPase in human cell line A-431 using PMCA4 ATPase Polyclonal Antibody (Product # PA5-83500). Staining shows localization to plasma membrane. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PMCA4 ATPase Antibody in Immunocytochemistry (ICC/IF)
          PMCA4 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PMCA4 ATPase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PMCA4 ATPase Antibody
          PMCA4 ATPase Polyclonal Antibody

          Product Details

          PA5-83500

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000-1:2,500
          -

          Immunocytochemistry (ICC/IF)

          0.25-2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein corresponding to Human ATP2B4. Recombinant protein control fragment (Product #RP-95778).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          10 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 40% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2790653

          Product Specific Information

          Immunogen sequence: THPEFAIEEE LPRTPLLDEE EEENPDKASK FGTRVLLLDG EVTPYANTNN NAVDCNQVQL PQSDSSLQSL ETSV

          Highest antigen sequence indentity to the following orthologs: Rat - 46%, Mouse - 53%.

          Target Information

          The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/K+ pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin (Cat. # MA3-917 and MA3-918).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATPase, Ca++ transporting, plasma membrane 4; Matrix-remodeling-associated protein 1; Plasma membrane calcium ATPase isoform 4; Plasma membrane calcium pump isoform 4; plasma membrane calcium-ATPase isoform 4 alternative splice variant x at splice site A; plasma membrane calcium-ATPase isoform 4, splice variant b at alternative splice site C; Plasma membrane calcium-transporting ATPase 4; PMCA4; sarcolemmal calcium pump; unnamed protein product

          View more View less

          Gene Aliases: ATP2B2; ATP2B4; MXRA1; PMCA4; PMCA4b; PMCA4x

          View more View less

          UniProt ID: (Human) P23634

          View more View less

          Entrez Gene ID: (Human) 493

          View more View less

          Function(s)
          nucleotide binding calcium-transporting ATPase activity protein binding calmodulin binding ATP binding calcium ion transmembrane transporter activity ATPase activity sodium channel regulator activity protein kinase binding PDZ domain binding protein phosphatase 2B binding nitric-oxide synthase inhibitor activity metal ion binding nitric-oxide synthase binding scaffold protein binding
          Process(es)
          neural retina development regulation of transcription from RNA polymerase II promoter ion transport calcium ion transport cellular calcium ion homeostasis spermatogenesis negative regulation of gene expression urinary bladder smooth muscle contraction negative regulation of angiogenesis hippocampus development sperm motility ion transmembrane transport nitric oxide-cGMP-mediated signaling pathway negative regulation of blood vessel endothelial cell migration negative regulation of nitric oxide biosynthetic process regulation of cytosolic calcium ion concentration response to hydrostatic pressure calcium ion transmembrane transport negative regulation of calcineurin-NFAT signaling cascade cellular response to epinephrine stimulus negative regulation of adrenergic receptor signaling pathway calcium ion transmembrane import into cytosol calcium ion import across plasma membrane negative regulation of the force of heart contraction transport across blood-brain barrier negative regulation of arginine catabolic process calcium ion export regulation of sodium ion transmembrane transport negative regulation of cellular response to vascular endothelial growth factor stimulus regulation of cell cycle G1/S phase transition positive regulation of protein localization to plasma membrane negative regulation of cardiac muscle hypertrophy in response to stress negative regulation of citrulline biosynthetic process regulation of cardiac conduction cellular response to acetylcholine
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline