Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Keep as concentrated solution.
Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (98%), Pig (100%), Chicken (100%), Sheep (100%), Rhesus Monkey (100%).
Positive Control: MCF-7, MDA-MB-231, mouse brain, rat brain.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ARF6 (ADP-ribosylation factor 6) is a broadly expressed, differentially regulated GTPase that is present in cytoplasm and on both cell-surface and intracellular membranes and whose functions may include tissue-specific effects on vesicular trafficking during cellular differentiation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP ribosylation factor 6; ADP-ribosylation factor 6; ARF6; DKFZp564M0264; small GTP binding protein; unnamed protein product
Gene Aliases: AI788669; ARF6; AW496366
UniProt ID: (Human) P62330, (Rat) P62332, (Mouse) P62331
Entrez Gene ID: (Human) 382, (Rat) 79121, (Mouse) 11845
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support