Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KHVKGILLYG PPGCGKTLLA RQIGKMLNAR EPKVVNGPEI LNKYVGESEA NIRKLFADAE EEQRRLGANS GLHIIIFDEI DAICKQRGSM AGSTGVHDTV VNQLLSKIDG VEQLNNILVI GMTNRPDLID EALLRPGRLE VKMEIGLPDE
Positive Samples: Mouse brain, Rat brain; Cellular Location: Cytoplasm
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NEM-sensitive fusion protein; Protein SKD2; Suppressor of K(+) transport growth defect 2; unnamed protein product; Vesicle-fusing ATPase; Vesicular-fusion protein NSF
Gene Aliases: AI316878; AU020090; AU067812; DEE96; Erg1; NSF; SEC18; SKD2
UniProt ID: (Human) A8K2D9, (Mouse) P46460, (Rat) Q9QUL6
Entrez Gene ID: (Human) 4905, (Mouse) 18195, (Rat) 60355
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support