Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: U-87MG; Cellular Location: Membrane, Multi-pass membrane protein
Immunogen sequence: SISFYGGFPF WGGLWFIISG SLSVAAENQP YSYCLLSGSL GLNIVSAICS AVGVILFITD LSIPHPYAYP DYYPYAWGVN PGMAISGVLL VFCLLEFGIA C
May be involved in signal transduction as a component of a multimeric receptor complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4SPAN4; Four-span transmembrane protein 4; Membrane-spanning 4-domains subfamily A member 8; Membrane-spanning 4-domains subfamily A member 8B; membrane-spanning 4-domains, subfamily A, member 8B; unnamed protein product
Gene Aliases: 4SPAN4; CD20L5; MS4A4; MS4A8; MS4A8B
UniProt ID: (Human) Q9BY19
Entrez Gene ID: (Human) 83661
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support