Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Positive Samples: A375, HT-29, U-251MG, LO2, Mouse heart, Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein
Immunogen sequence: SDKLYQRKEP VISSVHTKVK GIAEVKEEIV ENGVKKLVHS VFDTADYTFP LQGNSFFVMT NFLKTEGQEQ RLCPEYPTRR TLCSSDRGCK KGWMDPQSKG IQTGRCVVYE GNQKTCEVSA WCPIEAVEEA PRPALLNSAE NFTVLIKNNI DFPGHNYTTR NILPGLNITC TFHKTQNPQC PIFRLGDIFR ETGDNFSDVA IQGGIMGIEI YWDCNLDRWF HHCRPKYSFR RLDDKTTNVS LYPGYNFRYA KYYKENNVEK RTLIKVFGIR FDILVFGTGG KFDIIQLV
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and is responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Activation of this nuclear receptor by ATP in the cytoplasm may be a mechanism by which cellular activity can be coupled to changes in gene expression. Multiple alternatively spliced variants have been identified, most of which fit nonsense-mediated decay criteria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP agonist non-selective cationic pore-forming protein; ATP receptor; ligand gated ion channel; MGC20089; P2X purinoceptor 7; p2X receptor 7; P2X7; P2X7 purinoceptor; P2X7 receptor; P2X7; cervical cancer susceptibility protein; P2Z; P2Z receptor; Purinergic receptor; purinergic receptor P2X, ligand gated ion channel, 7; purinergic receptor P2X7; purnergic receptor P2X 7; unnamed protein product
Gene Aliases: AI467586; P2RX7; P2X(7); P2X7; P2X7R
UniProt ID: (Human) Q99572, (Rat) Q64663, (Mouse) Q9Z1M0
Entrez Gene ID: (Human) 5027, (Rat) 29665, (Mouse) 18439
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support