Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: SLGSKGFYYK DPSIPRLAKE PLAAAEPGLF GLNSGGHLQR AGEAERPSLH QRDGEMGAGR QQNPCPLFLG QPDTVPWTSW PACPPGLVHT LGNVWAGPGD GNLGYQLGPP ATPRCPSPEP PVTQRGCCSS YPPTKGGGLG PCGKCQEGLE GGASGASEPS EEVNKASGPR ACPPSHHTKL KKTWLTRHSE QFECPRGCPE VEERPVARLR ALKRAGSPEV QGAMGSPAPK RPPDPFPGTA EQGAGGWQEV RDTSIGNKDV DSGQHDEQKG PQDGQASLQD P; Positive Samples: U-251MG, Mouse brain, Rat brain; Cellular Location: Nucleus
Drosophila genes can be categorized based on the type of protein for which they encode and are represented by six major classifications, which include intracellular signaling proteins, transmembrane proteins, RNA binding proteins, secreted factors, transcription regulators (basic helix-loop-helix, homeodomain containing, zinc finger containing and chromatin associated) or other functional proteins. Among these numerous proteins, Hairless (CG5460, CT36411, CT17282) is a transcriptional co-repressor that contains a "Paired box" domain, a KX-repeat domain, a NEDL-repeat domain and a serine-rich domain. Hairless is an antagonist of the Notch signaling pathway in Drosophila.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-dimethyl-L-lysine(9) demethylase hairless; HAIR; hair growth associated; hairless homolog; Hairless protein; Lysine-specific demethylase hairless; protein hairless; restricted expression in the brain and skin
Gene Aliases: ALUNC; AU; ba; bldy; HR; HSA277165; HYPT4; MUHH; MUHH1; N; rh; rh-bmh; rhino
UniProt ID: (Human) O43593, (Mouse) Q61645
Entrez Gene ID: (Human) 55806, (Mouse) 15460, (Rat) 60563
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support