Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: VDEGEECSDS EGSHNEEAKE PDKTNKKEGE KTDRVGVASE ETPQTSSSSA RPGTPSDHQS QEASQFERKD EPKTEQTEKA EEETRMENSL PAKVPSRGDE MVADVQQPLS QIPSDTASPL LILPPPVPNP SPTLRPVETP VVGAPGMGSV S; Positive Samples: 293T, HeLa, Jurkat, Raji, LO2
The function of this protein remains unknown.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GIGYF2; GRB10 interacting GYF protein 2; GRB10-interacting GYF protein 2; Parkinson disease (autosomal recessive, early onset) 11; PERQ amino acid rich, with GYF domain 3; PERQ amino acid-rich with GYF domain-containing protein 2; rCG23949-like; trinucleotide repeat containing 15; Trinucleotide repeat-containing gene 15 protein; unnamed protein product
Gene Aliases: 2610016F01Rik; A830080H02Rik; AI852361; AW259676; BC006835; GIGYF2; GYF2; KIAA0642; mKIAA0642; PARK11; PERQ2; PERQ3; TNRC15
UniProt ID: (Human) Q6Y7W6, (Mouse) Q6Y7W8
Entrez Gene ID: (Human) 26058, (Mouse) 227331, (Rat) 100362458
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support