Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: YMKSKRREKK SSLLLLYKDE ERLWGEAMKP LPVVSGLRSV QVPLMLNMLQ ESVAPALSCT LCSMEGDSVS SESSSPDVHL TIQEEGADDE LEETSETPLN ESSEGSSENI HQNS; Positive Samples: 293T, LO2; Cellular Location: Membrane, Single-pass membrane protein
Ancillary protein that assembles as a beta subunit with a voltage-gated potassium channel complex of pore-forming alpha subunits. Modulates the gating kinetics and enhances stability of the channel complex. May associate with KCNQ1/KVLTQ1 and inhibit potassium current.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac voltage-gated potassium channel accessory subunit 4; Minimum potassium ion channel-related peptide 3; MINK-related peptide 3; MIRP3; MiRP3; Isk-related subfamily; Potassium channel subunit beta MiRP3; potassium channel, voltage gated subfamily E regulatory beta subunit 4; Potassium voltage-gated channel subfamily E member 4; potassium voltage-gated channel, Isk-related family, member 4; unnamed protein product
Gene Aliases: KCNE4; MIRP3
UniProt ID: (Human) Q8WWG9
Entrez Gene ID: (Human) 23704
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support