Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: PPVTVPGSTA TQKLLRTDKL EVCREFQRGN CARGETDCRF AHPADSTMID TSDNTVTVCM DYIKGRCMRE KCKYFHPPAH LQAKIKAAQH QANQAAVAAQ A; Positive Samples: A-549, HeLa, Mouse brain; Cellular Location: Cytoplasm, Nucleus
This gene encodes a C3H-type zinc finger protein, which is similar to the Drosophila melanogaster muscle blind B protein. Drosophila muscle blind is a gene required for photoreceptor differentiation. Several alternatively spliced transcript variants have been described but the full-length natures of only some have been determined.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3H-type zinc finger protein; DKFZp781H1296; MGC120625; MGC120626; MGC120628; muscleblind-like 2; Muscleblind-like protein 1; Muscleblind-like protein 2; Muscleblind-like protein-like; Muscleblind-like protein-like 39; predicted protein of HQ2032; RP11-128N14.1; unnamed protein product; zinc knuckle protein
Gene Aliases: 1110002M11Rik; AI047808; AI837313; AI849185; AL118326; Kiaa4072; MBLL; MBLL39; MBNL2; mKIAA4072; MLP1; PRO2032; R75232
UniProt ID: (Human) Q5VZF2, (Mouse) Q8C181
Entrez Gene ID: (Human) 10150, (Mouse) 105559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support