Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: DSAMNITAYL GNTTDNFFKI SNLKMGHNYT FTVQARCLFG NQICGEPAIL LYDELGSGAD ASATQAARST DVAAVVVPIL FLILLSLGVG FAILYTKHRR LQSSFTAFAN SHYSSRLGSA IFSSGDDLGE DDEDAPMITG FSDDVPMVIA; Positive Samples: SH-SY5Y, HepG2; Cellular Location: Endosome, Golgi apparatus, Membrane, Single-pass type I membrane protein
This gene encodes a mosaic protein that belongs to at least two families: the vacuolar protein sorting 10 domain-containing receptor family, and the low density lipoprotein receptor family. The encoded protein also contains fibronectin type III repeats and an epidermal growth factor repeat. The encoded protein is translated as a preproprotein and likely plays roles in endocytosis and sorting. There may be an association between expression of this locus and Alzheimer's Disease.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ21930; FLJ39258; LDLR relative with 11 ligand-binding repeats; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; mosaic protein LR11; SorLA; SorLA-1; Sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; Sorting protein-related receptor containing LDLR class A repeats; unnamed protein product
Gene Aliases: C11orf32; gp250; LR11; LRP9; SORL1; SORLA; SorLA-1
UniProt ID: (Human) Q92673
Entrez Gene ID: (Human) 6653
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support