Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MANGTADVRK LFIFTTTQNY FGLMSELWDQ PLLCNCLEIN NFLDDGNQML LRVQRSDAGI SFSNTIEFGD TKDKVLVFFK LRPEVITDEN LHDNILVSSM LESPISSLYQ AVRQVFAPML LKDQEWSRNF DPKLQNLLSE LEAGLGIVLR RSDTNLTKLK FKEDDTRGIL TPSDEFQFWI EQAHRGNKQI SKERANYFKE LFETIAREFY NLDSLSLLEV VDLVETTQDV VDDVWRQTEH DHYPESRMLH; Positive Samples: Rat testis; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein, cilium axoneme, cytoskeleton
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic dynein 2 heavy chain; Cytoplasmic dynein 2 heavy chain 1; cytoplasmic dynein heavy chain 2; Dynein cytoplasmic heavy chain 2; Dynein heavy chain isotype 1B; dynein, cytoplasmic, heavy chain 2; dynein, cytoplasmic, heavy polypeptide 2; Dynein-like protein 4
Gene Aliases: Dhc1b; Dlp4; Dnch2; Dnchc2; Dync2h1
UniProt ID: (Rat) Q9JJ79
Entrez Gene ID: (Rat) 65209
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support