Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MDSEKKRFTE EATKYFRERV SPVHLQILLT NNEAWKRFVT AAELPRDEAD ALYEALKKLR TYAAIEDEYV QQKDEQFREW FLKEFPQVKR KIQESIEKLR ALANGIEEVH RGCTISNVVS SSTGAASGIM SLAGLVLAPF TAGTSLALTA AGVGLGAASA VTGITTSIVE HSYTSSAEAE ASRLTATSID RLKVFKEVMR DITPNLLSLL NNYYEATQTI GSEIRAIRQA RARARLPVTT WRISAGSGGQ AERTIAGTTR AVSRGARILS ATTSGIFLAL DVVNLVYESK HLHEGAKSAS AEELRRQAQE LEENLMELTQ IYQRLNPCHT H; Positive Samples: DU145, A-549, BxPC-3, Mouse kidney; Cellular Location: Cytoplasm
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein L gene family encodes six highly homologous proteins designated apoL1-6, which are associated with large high density type lipoproteins (HDL). The human apoL family maps to chromosome 22q12.1-13.1 within a 127,000 bp region. apoL2 and apoL3 may allow the binding of lipids to organelles or may be involved in the movement of lipids in the cytoplasm. apoL2 localizes to the cytoplasm and is widely expressed, with the highest levels observed in the lung, thymus, pancreas, placenta, adult brain and prostate. The apoL2 protein is also detected in the spleen, liver, kidney, colon, small intestine, uterus, spinal cord, adrenal gland, salivary gland, trachea, mammary gland, skeletal muscle, testis and fetal brain and liver. apoL3 also localizes to the cytoplasm and is widely expressed, with highest levels detected in the prostate, lung and placenta. It is also detected in kidney, bone marrow, spleen, thymus, spinal cord, adrenal gland, salivary gland, trachea and mammary gland. Lower levels of apoL3 are observed in the brain, heart, fetal liver, pancreas and testis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ApoL-III; APOL3; apolipoprotein; apolipoprotein L, 3; Apolipoprotein L-III; Apolipoprotein L3; CG12-1; CG12_1; OTTHUMP00000028913; TNF-inducible protein CG12-1
Gene Aliases: apoL-III; APOL3; APOLIII; CG121; CG12_1
UniProt ID: (Human) O95236
Entrez Gene ID: (Human) 80833
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support