Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MAGIELERCQ QQANEVTEIM RNNFGKVLER GVKLAELQQR SDQLLDMSST FNKTTQNLAQ KKCWENIRYR ICVGLVVVGV LLIILIVLLV VFLPQSSDSS SAPRTQDAGI ASGPGN; Positive Samples: Mouse spleen; Cellular Location: Cell membrane, Endomembrane system, Golgi apparatus, Single-pass type IV membrane protein, trans-Golgi network membrane
Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of vesicles and cell membranes. The VAMP5 gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. This VAMP family member may participate in vesicle trafficking events that are associated with myogenesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myobrevin; vamp; VAMP-5; Vesicle-associated membrane protein 5
Gene Aliases: AF119384; Camp; Vamp5
UniProt ID: (Mouse) Q9Z2P8
Entrez Gene ID: (Mouse) 53620
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support