Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MQVPQDGEDL AGQPWYHGLL SRQKAEALLQ QNGDFLVRAS GSRGGNPVIS CRWRGSALHF EVFRVALRPR PGRPTALFQL EDEQFPSIPA LVHSYMTGRR PLSQATGAVV SRPVTWQGPL RRSFSEDTLM DGPARIEPLR ARKWSNSQPA DLAHMGRSRE DPAGMEASTM PISALPRTSS DPVLLKAPAP LGTVADSLRA SDGQLQAKAP TKPPRTPSFE LPDASERPPT YCELVPRVPS VQGTSPSQSC PEPEAPWWEA EEDEEEENRC FTRPQAEISF CPHDAPSCLL GPQNRPLEPQ; Positive Samples: BxPC-3, HeLa
May play a role in JNK activation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Novel SH2-containing protein 1; SH2 domain-containing protein 3A; unnamed protein product
Gene Aliases: NSP1; SH2D3A; UNQ175/PRO201
UniProt ID: (Human) Q9BRG2
Entrez Gene ID: (Human) 10045
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support