Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MEPVPLQDFV RALDPASLPR VLRVCSGVYF EGSIYEISGN ECCLSTGDLI KVTQVRLQKV VCENPKTSQT MELAPNFQGY FTPLNTPQSY ETLEELVSAT TQSSKQLPTC FMSTHRIVTE GRVVTEDQLL MLEAVVMHLG IRSARCVLGM EGQQVILHLP LSQKGPFWTW EPSAPRTLLQ VLQDPALKDL VLTCPTLPWH SLILRPQYEI QAIMHIFSSL RIAATRSAAQ TQGEDLARVH QGWLQYVQQD SCPQEGPQAR; Positive Samples: 293T, MCF7, Mouse spleen
May constitute a control point in macrophage inflammatory response, promoting LPS-induced TNF production.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basement membrane-induced; basement membrane-induced protein; Induced by contact to basement membrane 1 protein; Protein ICB-1; Protein THEMIS2; Thymocyte-expressed molecule involved in selection protein 2; unnamed protein product
Gene Aliases: C1orf38; ICB-1; ICB1; RGD1561016; THEMIS2
UniProt ID: (Human) Q5TEJ8, (Mouse) Q91YX0
Entrez Gene ID: (Human) 9473, (Rat) 500561, (Mouse) 230787
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support