Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: EAVGLQDIAP SPAAFPNFED STLFGREYID HLKAWLVSSG CPLQVKKAEP EPMREEEKMI PPTKPEIQAK APSSLSDAVP QRADHRVVGT IDQLVKRVIE GSLSPKERTL LKEDPAYWFL SDENSLEYKY YKLKLAEMQR MSENLRGADQ KPTSADCAVR AMLYSRAVRN LKKKLLPWQR RGLLRAQGLR GWKARRATTG T; Positive Samples: U-251MG, 293T, LO2, Jurkat, HeLa, Mouse thymus; Cellular Location: Nucleus
This gene encodes a member of the arginine/serine-rich family of splicing factors. The encoded protein functions in mRNA processing. Alternatively spliced transcript variants have been described. [provided by RefSeq].
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arginine/serine-rich-splicing factor 14; putative splicing factor, arginine/serine-rich 14; serine/arginine-rich splicing factor 14; Splicing factor, arginine/serine-rich 14; SURP and G-patch domain-containing protein 2; unnamed protein product
Gene Aliases: KIAA0365; mKIAA0365; SFRS14; SRFS14; Srsf14; SUGP2
UniProt ID: (Human) Q8IX01, (Mouse) Q8CH09
Entrez Gene ID: (Human) 10147, (Mouse) 234373
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support