Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MARGLQVPLP RLATGLLLLL SVQPWAESGK VLVVPTDGSP WLSMREALRE LHARGHQAVV LTPEVNMHIK EEKFFTLTAY AVPWTQKEFD RVTLGYTQGF; Positive Samples: Mouse liver, Mouse stomach, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Microsome, Single-pass membrane protein
This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. This enzyme has some glucuronidase activity towards bilirubin, although is is more active on amines, steroids, and sapogenins.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bilirubin UDP-glucuronosyltransferase isozyme 2; Bilirubin-specific UDPGT isozyme 1; Bilirubin-specific UDPGT isozyme 2; hUG-BR2; UDP glucuronosyltransferase 1 family, polypeptide A4; UDP glycosyltransferase 1 family, polypeptide A4; UDP-glucuronosyltransferase 1 family polypeptide A4s; UDP-glucuronosyltransferase 1-1; UDP-glucuronosyltransferase 1-4; UDP-glucuronosyltransferase 1-A; UDP-glucuronosyltransferase 1-D; UDP-glucuronosyltransferase 1A1; UDP-glucuronosyltransferase 1A4; UDPGT 1-1; UDPGT 1-4; UGT-1D; UGT1A4; unnamed protein product
Gene Aliases: GNT1; hUG-BR1; HUG-BR2; UDPGT; UDPGT 1-4; UGT-1A; UGT-1D; UGT1; UGT1-01; UGT1-04; UGT1.1; UGT1.4; UGT1A; UGT1A1; UGT1A4; UGT1A4S; UGT1D
UniProt ID: (Human) P22310
Entrez Gene ID: (Human) 54657
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support