Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • IHH Antibodies

          Invitrogen

          IHH Polyclonal Antibody

          View all (13) IHH antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IHH Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          IHH Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IHH Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IHH Antibody (PA5-93107) in WB

          Western blot analysis of IHH in extracts of various cell lines. Samples were incubated with IHH Polyclonal antibody (Product # PA5-93107) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 60s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IHH Antibody in Western Blot (WB)
          IHH Polyclonal Antibody

          Product Details

          PA5-93107

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 202-411 of human IHH (NP_0021722)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.51 mg/mL

          Amount

          151 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2806627

          Product Specific Information

          Immunogen sequence: GCFPAGAQVR LESGARVALS AVRPGDRVLA MGEDGSPTFS DVLIFLDREP HRLRAFQVIE TQDPPRRLAL TPAHLLFTAD NHTEPAARFR ATFASHVQPG QYVLVAGVPG LQPARVAAVS THVALGAYAP LTKHGTLVVE DVVASCFAAV ADHHLAQLAF WPLRLFHSLA WGSWTPGEGV HWYPQLLYRL GRLLLEEGSF HPLGMSGAGS; Positive Samples: Human placenta, Mouse kidney, Mouse liver, Mouse ovary, Rat liver, Rat lung, Rat brain; Cellular Location: Cell membrane, Extracellular side, Lipid-anchor, extracellular space

          Target Information

          Ihh encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HHG-2; IHH; Indian hedgehog homolog; Indian hedgehog protein

          View more View less

          Gene Aliases: BDA1; HHG2; IHH

          View more View less

          UniProt ID: (Human) Q14623, (Mouse) P97812

          View more View less

          Entrez Gene ID: (Human) 3549, (Mouse) 16147, (Rat) 84399

          View more View less

          Function(s)
          patched binding calcium ion binding protein binding peptidase activity transferase activity hydrolase activity very-low-density lipoprotein particle binding metal ion binding cholesterol-protein transferase activity molecular_function
          Process(es)
          skeletal system development ossification patterning of blood vessels osteoblast differentiation in utero embryonic development cell fate specification morphogenesis of a branching structure vasculature development heart looping positive regulation of mesenchymal cell proliferation epithelial cell morphogenesis retinal pigment epithelium development proteoglycan metabolic process proteolysis cell communication smoothened signaling pathway cell-cell signaling multicellular organism development pattern specification process positive regulation of cell proliferation embryo development embryonic pattern specification negative regulation of signal transduction intein-mediated protein splicing cell differentiation vitelline membrane formation pancreas development positive regulation of chondrocyte differentiation positive regulation of collagen biosynthetic process negative regulation of T cell differentiation in thymus negative regulation of immature T cell proliferation in thymus positive regulation of T cell differentiation in thymus multicellular organism growth chondrocyte proliferation regulation of growth embryonic digit morphogenesis camera-type eye development negative regulation of apoptotic process bone resorption regulation of cell differentiation negative regulation of cell differentiation positive regulation of smoothened signaling pathway positive regulation of transcription from RNA polymerase II promoter positive regulation of alpha-beta T cell differentiation negative regulation of alpha-beta T cell differentiation negative regulation of eye pigmentation cell maturation embryonic digestive tract morphogenesis embryonic camera-type eye morphogenesis neuron development smooth muscle tissue development positive regulation of epithelial cell proliferation cartilage development camera-type eye photoreceptor cell fate commitment head morphogenesis somite development embryonic skeletal joint development epithelial cell-cell adhesion response to estradiol maternal process involved in female pregnancy chondrocyte differentiation chondrocyte differentiation involved in endochondral bone morphogenesis cell proliferation response to mechanical stimulus regulation of gene expression protein autoprocessing negative regulation of chondrocyte differentiation system development self proteolysis liver regeneration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline