Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 7
Immunogen sequence: FMRQILNSDT IGCCGDDNGL MEVEGAHTSR TMSINAAELK QLLQSKEESP ENLFLELEKL VLEHSKDDDN LDSLLDSVVG LKQMLESSGD PLPLSDQDVE PVLSAPESLQ NLFNNRTAYV LADVMDDQLK SMWFTPFQAE EIDTDLDLVK VDLIELSEKC CSDFDLHSEL ERSFLSEPSS PGRTKTTKGF KLGKHKHETF ITSSGKSEYI EPAKRAHVVP PPRGRGRGGF GQGIRPHDIF RQRKQNTSRP PSMHVDDFVA AESKEVVPQD GIPPPKRPLK VSQKISSRGG FSGNRGGRGA F; Positive Samples: K-562, COS7, HeLa, PC-3
KIAA1429 is required for N6-methyladenosine (m6A) methylation of mRNAs, a modification that plays a role in the efficiency of mRNA splicing and processing. It is also Involved in mRNA splicing regulation, probably via its function in m6A methylation. Diseases associated with KIAA1429 include Retinitis Pigmentosa 42.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP434I116; functional spliceosome-associated protein 121; Protein virilizer homolog; unnamed protein product; virilizer homolog
Gene Aliases: 4930422M05Rik; fSAP121; KIAA1429; mKIAA1429; MSTP054; VIRMA
UniProt ID: (Mouse) A2AIV2, (Human) Q69YN4
Entrez Gene ID: (Mouse) 66185, (Rat) 313061, (Human) 25962
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support