Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Positive Samples: A-431, Mouse heart, Mouse brain, Rat heart, Rat brain; Cellular Location: Mitochondrion matrix
Immunogen sequence: KISLENVGCA SQIGKRKENE DRFDFAQLTD EVLYFAVYDG HGGPAAADFC HTHMEKCIMD LLPKEKNLET LLTLAFLEID KAFSSHARLS ADATLLTSGT TATVALLRDG IELVVASVGD SRAILCRKGK PMKLTIDHTP ERKDEKERIK KCGGFVAWNS LGQPHVNGRL AMTRSIGDLD LKTSGVIAEP ETKRIKLHHA DDSFLVLTTD GINFMVNSQE ICDFVNQCHD PNEAAHAVTE QAIQYGTEDN STAVVVPFGA WGK
PPM1K regulates the mitochondrial permeability transition pore and is essential for cellular survival and development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [3-methyl-2-oxobutanoate dehydrogenase (2-methylpropanoyl-transferring)]-phosphatase; [3-methyl-2-oxobutanoate dehydrogenase (2-methylpropanoyl-transferring)]-phosphatase, mitochondrial; BCKDH; Branched-chain alpha-ketoacid dehydrogenase phosphatase; branched-chain I+/--ketoacid dehydrogenase phosphatase; hPTMP; PP2C domain-containing protein phosphatase 1K; PP2C kappa; PP2C phosphatase; PP2C-kappa; PP2C-like mitochondrial protein; PP2C-like protein; PP2C-type mitochondrial phosphoprotein phosphatase; protein phosphatase 1K (PP2C domain containing); protein phosphatase 1K, mitochondrial; Protein phosphatase 2C family member; Protein phosphatase 2C isoform kappa; protein phosphatase 2C kappa; Protein phosphatase Mn(2+)-dependent 1K; PTMP; unnamed protein product
Gene Aliases: 2900063A19Rik; A930026L03Rik; BCKDH; BDP; MSUDMV; PP2Ckappa; PP2CM; PPM1K; PTMP; RGD1308501; UG0882E07
UniProt ID: (Human) Q8N3J5, (Rat) A6K136, (Mouse) Q8BXN7
Entrez Gene ID: (Human) 152926, (Rat) 312381, (Mouse) 243382
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support