Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MTIGRMENVE VFTAEGKGRG LKATKEFWAA DIIFAERAYS AVVFDSLVNF VCHTCFKRQE KLHRCGQCKF AHYCDRTCQK DAWLNHKNEC SAIKRYGKVP NENIRLAARI MWRVEREGTG LTEGCLVSVD DLQNHVEHFG EEEQKDLRVD VDTFLQYWPP QSQQFSMQYI SHIFGVINCN GFTLSDQRGL QAVGVGIFPN LGLVNHDCWP NCTVIFNNGN HEAVKSMFHT QMRIELRALG KISEGEELTV SYIDFLNVSE ERKRQLKKQY YFDCTCEHCQ KKLKDDLFLG VKDNPKPSQE VVKEMIQFSK DTLEKIDKAR SEGLYHEVVK LCRECLEKQE PVFADTNIYM; Positive Samples: Raji, MCF7, Mouse stomach, Rat heart, Rat stomach; Cellular Location: Cytoplasm, Nucleus
Acts as a transcriptional repressor. Essential for cardiomyocyte differentiation and cardiac morphogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD8 beta opposite; CD8b-opposite; CD8beta opposite strand; Histone-lysine N-methyltransferase SMYD1; m-BOP; SET and MYND domain-containing protein 1; unnamed protein product; Zinc finger protein BOP; zinc finger, MYND domain containing 18
Gene Aliases: 4632404M21Rik; BOP; C78565; KMT3D; SMYD1; ZMYND18; ZMYND22
UniProt ID: (Human) Q8NB12, (Mouse) P97443
Entrez Gene ID: (Human) 150572, (Mouse) 12180, (Rat) 297333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support