Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LGCLIYEMIE GQSPFRGRKE KVKREEVDRR VLETEEVYSH KFSEEAKSIC KMLLTKDAKQ RLGCQEEGAA EVKRHPFFRN MNFKRLEAGM LDPPFVPDPR AVYCKDVLDI EQFSTVKGVN LDHTDDDFYS KFSTGSVSIP WQNEMIETEC FKELNVFGPN GTLPPDLNRN HPPEPPKKGL LQRLFKRQHQ NNSKSSPSSK TSFNHHINSN HVSSNSTGSS; Positive Samples: A-549, U-251MG, HepG2, 293T, HT-1080, Mouse liver, Mouse lung; Cellular Location: Cell membrane, Cytoplasm, Nucleus, Peripheral membrane protein
GRK5 is a serine/threonine kinase that regulates the activity of a variety of G protein-coupled receptors and the motility of polymorphonuclear leukocytes. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-CP4; G protein-coupled receptor kinase 5; G protein-coupled receptor kinase GRK5; GRK; GRK-pan; Pan-GRK; Poly(rC)-binding protein 4; unnamed protein product
Gene Aliases: 1200003L19Rik; AlphaCP-4; FP2025; GPRK5; GRK5; Pcbp4
UniProt ID: (Human) P34947, (Mouse) Q8VEB1, (Rat) Q62833, (Mouse) P57724
Entrez Gene ID: (Human) 2869, (Mouse) 14773, (Rat) 59075, (Mouse) 59092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support