Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LERVKELQAE QEDRALRSFK LSVTVDPKYH PKIIGRKGAV ITQIRLEHDV NIQFPDKDDG NQPQDQITIT GYEKNTEAAR DAILRIVGEL EQMVSEDVPL DHRVHARIIG ARGKAIRKIM DEFKVDIRFP QSGAPDPNCV TVTGLPENVE EAIDHILNLE EEYLADVVDS EALQVYMKPP AHEEAKAPSR GFVVRDAPWT ASSSEKAPDM SSSEEFPSFG AQVAPKTLPW GPKR; Positive Samples: HeLa, Jurkat, HepG2, Mouse brain, NIH/3T3, Mouse heart, Rat liver; Cellular Location: Cytoplasm, Nucleus
High density lipoprotein-binding protein, also known as vigilin, is a 110-kD protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HDL-binding protein; High density lipoprotein-binding protein; lipoprotein-binding protein; predicted protein of HQ2900; unnamed protein product; Vigilin
Gene Aliases: 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDLBP; PRO2900; VGL
UniProt ID: (Human) Q00341, (Mouse) Q8VDJ3
Entrez Gene ID: (Human) 3069, (Mouse) 110611, (Rat) 64474
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support