Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen sequence: NLPSALVSHV LNPQPGEKIL DLCAAPGGKT THIAALMHDQ GEVIALDKIF NKVEKIKQNA LLLGLNSIRA FCFDGTKAVK LDMVEDTEGE PPFLPESFDR ILLDAPCSGM GQRPNMACTW SVKEVASYQP LQRKLFTAAV QLLKPEGVLV YSTCTITLAE NEEQVAWALT KFPCLQLQPQ EPQIGGEGMR GAGLSCEQLK QLQRFDPSAV PLPDTDMDSL REARREDMLR LANKDSIGFF IAKFVKCKST; Positive Samples: Jurkat, HepG2, HeLa, 293T, Mouse testis
NSun6 is an RNA methyltransferase that catalyzes the methylation at position cytosine C72 in some tRNAs. NSUN6 has been reported to localize to the cytoplasm and largely colocalize with Golgi apparatus and pericentriolar matrix, indicating that NSUN6 modifies tRNAs in a late step in their biogenesis. NSun6 may also play a major role in bone metastasis and could be a valuable therapeutic target for bone metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARL5B antisense RNA 1; NOL1/NOP2/Sun and PUA domain-containing protein 1; NOL1/NOP2/Sun domain family 6; NOL1/NOP2/Sun domain family member 6; NOL1/NOP2/Sun domain family, member 6; NOP2/Sun domain family, member 6; NOP2/Sun RNA methyltransferase family member 6; nucleolar protein (NOL1/NOP2/sun) and PUA domains 1; putative methyltransferase NSUN6; tRNA (cytosine(72)-C(5))-methyltransferase NSUN6; unnamed protein product
Gene Aliases: 4933403D21Rik; 4933414E04Rik; ARL5B-AS1; MRT82; NOPD1; NSUN6
UniProt ID: (Human) Q8TEA1, (Mouse) Q7TS68
Entrez Gene ID: (Human) 221078, (Mouse) 74455
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support