Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Immunogen sequence: TKKILMGNEE LTRLWNLCPD NMEACKSETR EHMPTLEEFF EEAIEQADPE NMVENEYKAV NNSNYGWRAL RLLARRSPHF FQPTNQQFKS LPEYLENMVI KLAKELPPPS EEIKTGEDED EEDNDALLKE NESPDVRRDK PVTGEQIEVF ANKLGEQWKI LAPYLEMKDS EIRQIECDSE DMKMRAKQLL VAWQDQEGVH ATPENLINAL NKSGLSDLAE SLTNDNETNS; Positive Samples: BT-474, MCF7, A-549, HeLa, HepG2; Cellular Location: Cytoplasm, Cytoplasm, Nucleus, Nucleus matrix, Nucleus speckle, nucleoplasm
HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hTREX84; Nuclear matrix protein p84; p84N5; p84N5s; p84N5s2; THO complex 1; THO complex subunit 1; tho1; unnamed protein product
Gene Aliases: 3110002N20Rik; AW107452; DFNA86; HPR1; LRRGT00070; NMP-84; P84; P84N5; THOC1
UniProt ID: (Human) Q96FV9, (Mouse) Q8R3N6
Entrez Gene ID: (Human) 9984, (Mouse) 225160, (Rat) 291797
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support