Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: SH-SY5Y, Mouse brain
Immunogen sequence: QPEAAQQEAV TAAEHPGLDD FLRQVERLLF LRENIQRLQG DQGEHSASQI FQSDWLSKRQ HPGKREEEEE EGVEEEEEEE GGAVGPHKRQ HPGRREDEAS WSVDVTQHKR QHPGRRSPWL AYAVPKRQHP GRRLADPKAQ RSWEEEEEEE EREEDLMPEK RQHPGKRALG GPCGPQGAYG QAGLLLGLLD DLSRSQGAEE KRQHPG
This gene encodes a member of the thyrotropin-releasing hormone family. Cleavage of the encoded proprotein releases mature thyrotropin-releasing hormone, which is a tripeptide hypothalamic regulatory hormone. The human proprotein contains six thyrotropin-releasing hormone tripeptides. Thyrotropin-releasing hormone is involved in the regulation and release of thyroid-stimulating hormone, as well as prolactin. Deficiency of this hormone has been associated with hypothalamic hypothyroidism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pro-thyrotropin-releasing hormone; Pro-TRH; Prothyroliberin; protirelin; thyrotropin releasing hormone; thyrotropin-releasing factor; TSH-releasing factor; unnamed protein product
Gene Aliases: Pro-TRH; TRF; TRH
UniProt ID: (Human) P20396, (Mouse) Q62361
Entrez Gene ID: (Human) 7200, (Mouse) 22044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support