Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • GATA1 Antibodies

          Invitrogen

          Phospho-GATA1 (Ser142) Polyclonal Antibody

          View all (29) GATA1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-GATA1 (Ser142) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (20)
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-GATA1 (Ser142) Antibody (PA5-99361) in IHC (P)

          Immunohistochemistry analysis of paraffin-embedded Phospho-GATA1 (Ser142) in human bladder cancer tissue sections. Antigen retrieval was performed using citrate buffer. Samples were blocked with blocking buffer (1.5 hr, 22°C), incubated with Phospho-GATA1 (Ser142) polyclonal antibody (Product # PA5-99361) using a dilution of 1:200 (1.5 hr, 22°C), followed by HRP conjugated goat anti-rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-GATA1 (Ser142) Antibody in Western Blot (WB)
          Phospho-GATA1 (Ser142) Polyclonal Antibody

          Product Details

          PA5-99361

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human GATA1(Accession P15976), corresponding to amino acid residues around phosphorylated Ser142.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2818294

          Product Specific Information

          Antibody detects endogenous levels of GATA1 only when phosphorylated at Serine 142.

          Target Information

          Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Eryf1; erythroid DNA-binding protein; Erythroid transcription factor; erythroid transcription factor 1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF-1; globin transcription factor 1; HGNC:4170; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; unnamed protein product

          View more View less

          Gene Aliases: CNSHA9; ERYF1; GATA-1; GATA1; GF-1; GF1; HAEADA; NF-E1; NFE1; XLANP; XLTDA; XLTT

          View more View less

          UniProt ID: (Human) P15976, (Mouse) P17679, (Rat) P43429

          View more View less

          Entrez Gene ID: (Human) 2623, (Mouse) 14460, (Rat) 25172

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding transcription cofactor binding transcription coactivator binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding p53 binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding zinc ion binding chromatin DNA binding sequence-specific DNA binding metal ion binding RNA polymerase II sequence-specific DNA binding transcription factor binding C2H2 zinc finger domain binding sequence-specific double-stranded DNA binding RNA polymerase II core promoter sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II transcription factor binding enhancer sequence-specific DNA binding DNA binding, bending DNA-binding transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter positive regulation of cytosolic calcium ion concentration cell-cell signaling cell proliferation negative regulation of cell proliferation male gonad development anatomical structure morphogenesis regulation of glycoprotein biosynthetic process regulation of definitive erythrocyte differentiation regulation of primitive erythrocyte differentiation myeloid cell differentiation cell differentiation erythrocyte differentiation megakaryocyte differentiation platelet formation basophil differentiation eosinophil differentiation bone mineralization negative regulation of bone mineralization organ regeneration myeloid cell apoptotic process negative regulation of myeloid cell apoptotic process osteoblast proliferation positive regulation of osteoblast proliferation embryonic hemopoiesis eosinophil fate commitment negative regulation of apoptotic process positive regulation of mast cell degranulation cell fate commitment positive regulation of erythrocyte differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cell development system development erythrocyte development homeostasis of number of cells within a tissue Sertoli cell development primitive erythrocyte differentiation platelet aggregation cellular response to lipopolysaccharide cellular response to cAMP cellular response to follicle-stimulating hormone stimulus dendritic cell differentiation negative regulation of extrinsic apoptotic signaling pathway in absence of ligand transcription, DNA-templated positive regulation of peptidyl-tyrosine phosphorylation transcriptional activation by promoter-enhancer looping cellular response to thyroid hormone stimulus negative regulation of transcription regulatory region DNA binding
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline