Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total PDE4B/C/D.
This gene is a member of the type IV, cyclic AMP (cAMP)-specific, cyclic nucleotide phosphodiesterase (PDE) family. The encoded protein regulates the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. Altered activity of this protein has been associated with schizophrenia and bipolar affective disorder. Alternative splicing and the use of alternative promoters results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3',5'-cyclic-AMP phosphodiesterase 4B; 3',5'-cyclic-AMP phosphodiesterase 4C; 3',5'-cyclic-AMP phosphodiesterase 4D; alternatively spliced; cAMP specific rolipram-sensitive phosphodiesterase; cAMP specific rolipram-sensitive phosphosdiesterase; cAMP-specific 3',5'-cyclic phosphodiesterase 4B; cAMP-specific 3',5'-cyclic phosphodiesterase 4C; cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase 4B; cAMP-specific phosphodiesterase 4C; cAMP-specific phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D6; cyclic AMP specific phosphodiesterase PDE4D5A; DPDE1; DPDE3; DPDE4; dunce; dunce-like phosphodiesterase E1; dunce-like phosphodiesterase E4; PDE21; PDE32; PDE43; PDE4B5; phosphodiesterase 4B, cAMP-specific (phosphodiesterase E4 dunce homolog, Drosophila); phosphodiesterase 4C, cAMP-specific (phosphodiesterase E1 dunce homolog, Drosophila); phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); testicular tissue protein Li 136; unnamed protein product
Gene Aliases: 9630011N22Rik; ACRDYS2; DPDE1; DPDE3; DPDE4; dunce; E130301F19Rik; HSPDE4D; PDE21; PDE43; PDE4B; PDE4C; PDE4D; PDE4DN2; PDEIVB; R74983; STRK1
UniProt ID: (Human) Q07343, (Human) Q08493, (Human) Q08499, (Mouse) Q3UEI1, (Mouse) Q01063
Entrez Gene ID: (Human) 5142, (Human) 5143, (Human) 5144, (Mouse) 18578, (Mouse) 110385, (Mouse) 238871
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support