Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total GOLT1A.
GOLT1A may be involved in fusion of ER-derived transport vesicles with the Golgi complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Golgi transport 1 homolog A; golt1a; hGOT1b; RP11-203F10.3; unnamed protein product; Vesicle transport protein GOT1A
Gene Aliases: 0610012C01Rik; CGI-141; GOLT1A; GOT1; GOT1B; hGOT1b; HMFN1187; Kiss1; metastatin; YMR292W
UniProt ID: (Human) Q6ZVE7, (Mouse) Q9DCQ3
Entrez Gene ID: (Human) 127845, (Mouse) 68338
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support