Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (400 µL) be added to the vial to prepare a stock solution of 0.25 mg/mL.
Concentration is measured by UV.
CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD64; CD64 antigen; Fc-gamma receptor 1; Fc-gamma RI; FcgRI; FcRI; FLJ18345; High affinity immunoglobulin gamma Fc receptor I; IgG Fc receptor I; IgG high affinity Fc receptor; RP11-196G18.2
Gene Aliases: AI323638; AV092959; CD64; Fcg1; FcgammaRI; Fcgr1; IGGHAFC
UniProt ID: (Mouse) P26151
Entrez Gene ID: (Mouse) 14129
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support