Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57138. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Required for organization and/or function of the secretory apparatus in Clara cells in lung.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hikeshi, heat shock protein nuclear import factor; lethal, Chr 7, Rinchik 6; unnamed protein product
Gene Aliases: C11orf73; HLD13; HSPC138; HSPC179; L7RN6; OPI10
UniProt ID: (Human) Q53FT3
Entrez Gene ID: (Human) 51501
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support