Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61649. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of DDX49 has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEAD (Asp-Glu-Ala-Asp) box polypeptide 49; DEAD box protein 49; Probable ATP-dependent RNA helicase DDX49; unnamed protein product
Gene Aliases: Dbp8; DDX49; R27090_2
UniProt ID: (Human) Q9Y6V7
Entrez Gene ID: (Human) 54555
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support