Search
Search

TAB2| Gene Symbol: TAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001292034.3 | NP_001278963.1 | ||||
| NM_001292035.3 | NP_001278964.1 | |||||
| NM_001369506.1 | NP_001356435.1 | |||||
| NM_015093.6 | NP_055908.1 | |||||
| NR_125861.2 | ||||||
| XM_011535633.3 | XP_011533935.1 | |||||
| XM_017010592.3 | XP_016866081.1 | |||||
| XM_047418485.1 | XP_047274441.1 | |||||
| XM_047418486.1 | XP_047274442.1 | |||||
| XM_047418487.1 | XP_047274443.1 | |||||
| XM_047418488.1 | XP_047274444.1 | |||||
| XM_047418489.1 | XP_047274445.1 | |||||
| XM_047418490.1 | XP_047274446.1 | |||||
| GenBank mRNA | AB018276.2 | - | ||||
| AF241230.1 | - | |||||
| AI159838.1 | - | |||||
| AK025538.1 | - | |||||
| AK295750.1 | - | |||||
| AK307650.1 | - | |||||
| AK315038.1 | - | |||||
| AL050180.1 | - | |||||
| AL117407.1 | - | |||||
| BC035910.2 | - | |||||
| BC041998.1 | - | |||||
| BM661637.1 | - | |||||
| BX645362.1 | - | |||||
| CR457387.1 | - | |||||
| DA154923.1 | - | |||||
| DB467477.1 | - | |||||