Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000586.3 | NP_000577.2 | ||||
XM_017008177.1 | XP_016863666.1 | |||||
GenBank mRNA | A14844.1 | - | ||||
AB451325.1 | - | |||||
AB451454.1 | - | |||||
AF228636.1 | - | |||||
AF532913.1 | - | |||||
AY283686.1 | - | |||||
AY523040.1 | - | |||||
BC066254.1 | - | 6971690 | ||||
BC066255.1 | - | 6971691 | ||||
BC066256.1 | - | 6971693 | ||||
BC066257.1 | - | 6971694 | ||||
BC070338.1 | - | 30418817 | ||||
DQ231169.1 | - | |||||
DQ861285.1 | - | |||||
ENST00000226730.0 | - | |||||
S77834.1 | - | |||||
S77835.1 | - | |||||
S82692.1 | - | |||||
U25676.1 | - | |||||
V00564.1 | - | |||||
X01586.1 | - |