Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203712.1 | NP_001190641.1 | 5 - 6 | 476 | 66 | |
NM_001345833.1 | NP_001318898.1 | 4 - 5 | 378 | 66 | ||
NM_126150.5 | NP_201551.1 | 4 - 5 | 463 | 66 | ||
GenBank mRNA | AY085100.1 | - | 4 - 5 | 375 | 66 | |
BT025802.1 | - | 4 - 5 | 315 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |