SuperScript™ VILO™ cDNA Synthesis Kit
Invitrogen™

SuperScript™ VILO™ cDNA Synthesis Kit

The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
1175405050 Reactions
11754250250 Reactions
Catalog number 11754050
Price (TWD)
23,040.00
Online offer
Ends: 30-Jun-2026
28,800.00
Save 5,760.00 (20%)
Each
No. of Reactions:
50 Reactions
Request bulk or custom format
Price (TWD)
23,040.00
Online offer
Ends: 30-Jun-2026
28,800.00
Save 5,760.00 (20%)
Each

The SuperScript VILO cDNA Synthesis Kit is designed to generate first strand cDNA for two-step RT-qPCR applications. The kit is supplied in a two-tube format with the VILO reaction mix and an enzyme blend in separate tubes. The enzyme blend contains SuperScript III Reverse Transcriptase (RT), a genetically engineered MMLV RT that has reduced RNase H activity and improved thermostability for highly efficient cDNA synthesis.

The SuperScript VILO cDNA Synthesis Kit can be used to synthesize cDNA from for a wide range of input RNA amounts. This kit provides the high temperature capability of SuperScript III Reverse Transcriptase in an optimized format for generating first-strand cDNA for use in RT–qPCR.

Notes
• For superior first-strand cDNA synthesis in two-step RT-qPCR, try SuperScript IV VILO Master Mix or SuperScript IV VILO Master Mix with ezDNase Enzyme.
• This kit is included with EXPRESS Two-Step RT–qPCR kits. Reagents are provided for 50 or 250 20-μL cDNA synthesis reactions.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
No. of Reactions50 Reactions
Optimal Reaction Temperature42°C
Quantity50 reactions
Reaction FormatMaster Mix
Reagent TypeReverse Transcription
Reverse TranscriptaseSuperScript III
Ribonuclease H ActivityReduced
Shipping ConditionDry Ice
Size (Final Product)Up to 10 kb
Starting MaterialRNA
TechniqueReverse Transcription
Detection MethodqPCR
For Use With (Application)Real Time PCR (qPCR)
GC-Rich PCR PerformanceHigh
Reaction Speed60 min.
Unit SizeEach
Contents & Storage

• 5X VILO Reaction Mix, 200 μL
• 10X SuperScript III Enzyme Blend, 100 μL

Store at –5 to –30°C.

Frequently asked questions (FAQs)

What is the difference between SuperScript III RT and the RT in the SuperScript VILO kit?

The SuperScript VILO cDNA Synthesis Kit contains a mix of SuperScript III RT and helper proteins which help to increase the efficiency of the reverse transcription reaction and thus improve yield. The RT in the SuperScript VILO kit is active at 42 degrees C due to the helper proteins.

How much of the first-strand cDNA reaction should I load for PCR?

While the volume is dependent on the starting amount of RNA used for the first-strand synthesis and the abundance of the target gene, we'd recommend starting with 10% of the first-strand reaction for your PCR reaction.

In what concentration range of total RNA does the SuperScript VILO cDNA Synthesis Kit (Cat. No. 11754050) work?

  • Lower limit of RNA concentration: 1.0 pg
  • Upper limit of RNA concentration: 2.5 µg


  • Find additional tips, troubleshooting help, and resources within our Reverse Transcription and RACE Support Center

    What is the RNA input for the Oncomine Focus Assay, Chef-Ready Library kit?

    Each reverse transcription reaction requires 10 ng of DNase-treated total RNA (≥1.43 ng/µL).

    With the Oncomine Focus Assay, Chef-Ready Library, can I perform reverse transcription using the SuperScript VILO cDNA Synthesis Kit on the Ion Chef System?

    Reverse transcription needs to be performed manually in the IonCode 96-well PCR plate, using the SuperScript VILO cDNA Synthesis Kit. SuperScript reverse transcriptase enzyme is not supplied in the kit and must be purchased separately. If the RNA was prepared from FFPE tissue and not previously heat-treated, we recommend heating at 80 degrees C for 10 min, then cooling to room temperature.

    Citations & References (5)

    Citations & References
    Abstract
    A useful approach to total analysis of RISC-associated RNA.
    Authors:Hayashida Y, Nishibu T, Inoue K, Kurokawa T,
    Journal:BMC Res Notes
    PubMed ID:19706194
    'ABSTRACT: BACKGROUND: Identifying the endogenous RNA induced silencing complex(RISC)-associated RNAs is essential for understanding the cellular regulatory networks by miRNAs. Recently, isolation of RISC-associated mRNAs using antibody was reported, but their method needs a large amount of initial materials. We tried to improve the protocol and constructed an efficient and ... More
    Transcriptome analysis of proliferating Arabidopsis endosperm reveals biological implications for the control of syncytial division, cytokinin signaling, and gene expression regulation.
    Authors:Day RC, Herridge RP, Ambrose BA, Macknight RC,
    Journal:Plant Physiol
    PubMed ID:18923020
    During the early stages of seed development, Arabidopsis (Arabidopsis thaliana) endosperm is syncytial and proliferates rapidly through repeated rounds of mitosis without cytokinesis. This stage of endosperm development is important in determining final seed size and is a model for studying aspects of cellular and molecular biology, such as the ... More
    T cells modulate Epstein-Barr virus latency phenotypes during infection of humanized mice.
    Authors:Heuts F, Rottenberg ME, Salamon D, Rasul E, Adori M, Klein G, Klein E, Nagy N,
    Journal:
    PubMed ID:24390326
    Human B cells, the main target of Epstein-Barr virus (EBV), can display several types of latent viral protein expression, denoted 0, I, IIa, IIb, or III. Of these, only type III expression induces proliferation of cells in vitro. These latency types are present at specific stages of infection and are ... More
    Decrease of miR-146b-5p in monocytes during obesity is associated with loss of the anti-inflammatory but not insulin signaling action of adiponectin.
    Authors:Hulsmans M, Van Dooren E, Mathieu C, Holvoet P,
    Journal:PLoS One
    PubMed ID:22393448
    Low adiponectin, a well-recognized antidiabetic adipokine, has been associated with obesity-related inflammation, oxidative stress and insulin resistance. Globular adiponectin is an important regulator of the interleukin-1 receptor-associated kinase (IRAK)/NF?B pathway in monocytes of obese subjects. It protects against inflammation and oxidative stress by inducing IRAK3. microRNA (miR)-146b-5p inhibits NF?B-mediated inflammation ... More
    Cyclin D1 splice variants: polymorphism, risk, and isoform-specific regulation in prostate cancer.
    Authors:Comstock CE, Augello MA, Benito RP, Karch J, Tran TH, Utama FE, Tindall EA, Wang Y, Burd CJ, Groh EM, Hoang HN, Giles GG, Severi G, Hayes VM, Henderson BE, Le Marchand L, Kolonel LN, Haiman CA, Baffa R, Gomella LG, Knudsen ES, Rui H, Henshall SM, Sutherland RL, Knudsen KE,
    Journal:Clin Cancer Res
    PubMed ID:19706803
    PURPOSE: Alternative CCND1 splicing results in cyclin D1b, which has specialized, protumorigenic functions in prostate not shared by the cyclin D1a (full length) isoform. Here, the frequency, tumor relevance, and mechanisms controlling cyclin D1b were challenged. EXPERIMENTAL DESIGN: First, relative expression of both cyclin D1 isoforms was determined in prostate ... More