Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ETL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126270.2 | NP_178318.1 | 10 - 11 | 2166 | 88 | |
GenBank mRNA | AY059857.1 | - | 10 - 11 | 2105 | 88 | |
BT002576.1 | - | 10 - 11 | 1908 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |