Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AT-IE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332983.1 | NP_001322576.1 | 2 - 3 | 1290 | 154 | |
NM_001332984.1 | NP_001322577.1 | 2 - 3 | 471 | 154 | ||
NM_102923.4 | NP_174469.1 | 2 - 3 | 471 | 154 | ||
GenBank mRNA | AB006082.1 | - | 2 - 3 | 439 | 154 | |
AY046000.1 | - | 2 - 3 | 465 | 154 | ||
AY079376.1 | - | 2 - 3 | 394 | 154 | ||
AY086514.1 | - | 2 - 3 | 471 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |