Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336987.1 | NP_001324501.1 | 3 - 4 | 1046 | 85 | |
NM_180041.5 | NP_850372.4 | 4 - 5 | 1028 | 85 | ||
NM_180042.5 | NP_850373.4 | 4 - 5 | 736 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |