Search
Search

AT3G53740| Gene Symbol: AT3G53740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035778.3 | NP_001030855.1 | 2 - 3 | 155 | 86 | |
| NM_001084810.2 | NP_001078279.1 | 1 - 2 | 243 | 86 | ||
| NM_115234.2 | NP_566987.1 | 2 - 3 | 175 | 86 | ||
| NM_180366.4 | NP_850697.1 | 2 - 3 | 233 | 86 | ||
| GenBank mRNA | AY062791.1 | - | 2 - 3 | 159 | 86 | |
| AY081579.1 | - | 1 - 2 | 109 | 86 | ||
| AY086677.1 | - | 2 - 3 | 176 | 86 | ||
Set Membership: |
|