Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337803.1 | NP_001326171.1 | 4 - 5 | 654 | 129 | |
NM_111750.4 | NP_566344.2 | 4 - 5 | 669 | 129 | ||
NM_180665.1 | NP_850996.1 | 4 - 5 | 594 | 129 | ||
NM_202535.1 | NP_974264.1 | 3 - 4 | 770 | 129 | ||
GenBank mRNA | AB045112.1 | - | 4 - 5 | 627 | 129 | |
AY099706.1 | - | 4 - 5 | 670 | 129 | ||
AY128900.1 | - | 4 - 5 | 532 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |