Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036960.2 | NP_001032037.1 | 5 - 6 | 445 | 93 | |
NM_001344808.1 | NP_001331840.1 | 5 - 6 | 617 | 93 | ||
NM_124265.5 | NP_568704.2 | 4 - 5 | 1312 | 93 | ||
NM_180826.4 | NP_851157.1 | 4 - 5 | 462 | 93 | ||
GenBank mRNA | AF062590.1 | - | 4 - 5 | 342 | 93 | |
AF062591.1 | - | 4 - 5 | 561 | 93 | ||
AY079026.1 | - | 4 - 5 | 447 | 93 | ||
AY136455.1 | - | 4 - 5 | 350 | 93 | ||
BT002594.1 | - | 3 - 4 | 249 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |