Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198391.1 | NP_001185320.1 | 5 - 6 | 1358 | 78 | |
NM_105221.3 | NP_176725.1 | 5 - 6 | 1409 | 78 | ||
GenBank mRNA | AY126994.1 | - | 5 - 6 | 1358 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |