Search Thermo Fisher Scientific
Gene Symbol: CYP81D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123013.4 | NP_568533.2 | 1 - 2 | 781 | 128 | |
GenBank mRNA | AY093766.1 | - | 1 - 2 | 529 | 128 | |
BT001045.1 | - | 1 - 2 | 512 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |